missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) phenylacetylide, Thermo Scientific Chemicals

Catalog No. p-7049656
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1753804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1753804 Supplier Thermo Scientific Chemicals Supplier No. 01753804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13146-23-1 | Molecular Formula: C8H5Cu | Molecular Weight: 164.674 g/mol

Copper(I) phenylacetylide participates in efficient coupling with a range of aryl iodides and bromides to give substituted tolanes which, if possessing an ortho amino or hydroxy group, can be thermolyzed to yield 2-phenylbenzofurans, -indoles, and -furo[3,2-b]pyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13146-23-1
Molecular Formula C8H5Cu
Molecular Weight (g/mol) 164.674
MDL Number MFCD00015578
InChI Key SFLVZUJLUPAJNE-UHFFFAOYSA-N
Synonym copper 1+ ; ethynylbenzene, copper i phenylacetylide, copper i phenylacetylide, min
PubChem CID 6101206
IUPAC Name copper(1+);ethynylbenzene
SMILES [C-]#CC1=CC=CC=C1.[Cu+]

Specifications

Color Yellow
Quantity 2 g
UN Number UN3088
Sensitivity Air and moisture sensitive
Solubility Information Soluble in acetone and acetonitrile; Insoluble in water
Formula Weight 153.68
Physical Form Powder
Chemical Name or Material Copper(I) phenylacetylide
Hazard Category H250-H260-H314-H335-X
Hazard Statement GHS H Statement
H251-H260-H314-H318
Self-heating: may catch fire.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P223-P231+P232-P235+P410-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: SELF-HEATING SOLID, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only