missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) cyanide

Catalog No. AA12135F3
Change view
Click to view available options
Quantity:
250 g
1 kg
2 x 2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12135F3 2 x 2.5 kg
AA1213530 250 g
AA12135A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA12135F3 Supplier Thermo Scientific Chemicals Supplier No. 012135F3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Electroplating Cu or Fe, Polymerization Catalyst | CAS: 544-92-3 | CCuN | 89.564 g/mol

Copper(I) cyanide is commonly used as a polymerization catalyst. The compound is useful in organic synthesis, as a catalyst, reagent in the preparation of nitriles(sandmeyer reaction). It is also used in electroplating copper and iron, silver plating, brass plating, and copper-tin alloy plating. It is used to make phthalocyanine dyes and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.564
MDL Number MFCD00010975
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]

Specifications

Melting Point 473°C
Color White
Physical Form Powder
Quantity 2 x 2.5 kg
Beilstein 3587244
Solubility Information Practically insoluble in water and alcohol. Soluble in ammonium hydroxide,aqueous ammonia,pyridine and N-methylpyrrolidone.
Formula Weight 89.56
Odor Characteristic
Density 2.92
Chemical Name or Material Copper(I) cyanide
Hazard Category H300+H310+H330
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 208-883-6
RTECSNumber GL7150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only