missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Cyanide 98.0+%, TCI America™

Catalog No. p-7125208
Change view
Click to view available options
Quantity:
25 g
300 g
Catalog No. Quantity
C195225G 25 g
C1952300G 300 g
2 options

Catalog No. C195225G

Supplier: TCI America C195225G

Only null left

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.564
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]

Specifications

Melting Point 473°C
Color Green-White
Physical Form Crystalline Powder
Quantity 25 g
UN Number 1587
Formula Weight 89.56
Percent Purity ≥98.0% (T)
Chemical Name or Material Copper(I) Cyanide
EINECSNumber (1)-0376
RTECSNumber GL7150000
TSCA Yes