missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Cyanide 98.0+%, TCI America™

Catalog No. C1952300G
Change view
Click to view available options
Quantity:
25 g
300 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1952300G 300 g
C195225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1952300G Supplier TCI America Supplier No. C1952300G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 544-92-3 | CCuN | 89.564 g/mol

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.564
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]

Specifications

Melting Point 473°C
Color Green-White
Physical Form Crystalline Powder
Quantity 300 g
UN Number 1587
Formula Weight 89.56
Percent Purity ≥98.0% (T)
Chemical Name or Material Copper(I) Cyanide
EINECSNumber (1)-0376
RTECSNumber GL7150000
TSCA Yes