missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 99%, extra pure, purified

Catalog No. p-222897
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208390250 25 g Glass bottle
AC208391000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC208390250 Supplier Thermo Scientific Chemicals Supplier No. 208390250
Only null left

CAS: 7758-89-6 | ClCu | 99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 99
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Boiling Point 1490.0°C
Melting Point 430.0°C
Color Gray-Green
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.5 to 101.5% (Complexometry)
Linear Formula CuCl
Fieser 01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97
Merck Index 15,265
Solubility Information Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh, forming complexes, practically insoluble in alcohol and acetone
Formula Weight 99
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 4.14
Density 4.1400g/mL
Chemical Name or Material Copper(I) chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
EINECSNumber 231-842-9
RTECSNumber GL6990000
TSCA TSCA

RUO – Research Use Only