Learn More
Copper(I) chloride, 99.999% (metals basis)
CAS: 7758-89-6 | ClCu | 98.996 g/mol
$361.89 - $361.89
Chemical Identifiers
| CAS | 7758-89-6 |
|---|---|
| Molecular Formula | ClCu |
| Molecular Weight (g/mol) | 98.996 |
| MDL Number | MFCD00010971 |
| InChI Key | OXBLHERUFWYNTN-UHFFFAOYSA-M |
| Synonym | cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech |
| PubChem CID | 62652 |
| ChEBI | CHEBI:53472 |
| IUPAC Name | chlorocopper |
| SMILES | Cl[Cu] |
Description
Copper(I) chloride is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7758-89-6 | |
| 98.996 | |
| OXBLHERUFWYNTN-UHFFFAOYSA-M | |
| 62652 | |
| chlorocopper |
| ClCu | |
| MFCD00010971 | |
| cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
| CHEBI:53472 | |
| Cl[Cu] |
Specifications
| 1490°C | |
| 7758-89-6 | |
| 25 g | |
| ClCu | |
| cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
| OXBLHERUFWYNTN-UHFFFAOYSA-M | |
| chlorocopper | |
| 62652 | |
| 98.99 | |
| Air and moisture sensitive | |
| Copper(I) chloride |
| 430°C | |
| Powder | |
| 99.999% (Metal basis) | |
| MFCD00010971 | |
| Very soluble in concentrated HCl. Soluble in ammonium hydroxide. Insoluble in ethanol and acetone. Sparingly soluble in water. | |
| Cl[Cu] | |
| 98.996 | |
| CHEBI:53472 | |
| 1.93 | |
| 4.14 |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H318
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-842-9
RTECSNumber : GL6990000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only