missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 97%

Catalog No. p-3256996
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1187122 100 g
AA1187136 500 g
AA11871A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1187122 Supplier Thermo Scientific Chemicals Supplier No. 01187122
Only null left

CAS: 7758-89-6 | ClCu | 98.996 g/mol

It is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 98.996
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Boiling Point 1490°C
Melting Point 430°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 97%
Solubility Information Very soluble in concentrated HCl. Soluble in ammonium hydroxide. Insoluble in ethanol and acetone. Sparingly soluble in water.
Formula Weight 98.99
Refractive Index 1.93
Sensitivity Air and moisture sensitive
Appearance Powder
Density 4.14 g/cm3
Chemical Name or Material Copper(I) chloride
Show More Show Less
Hazard Category H302+H312-H315-H318
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-842-9
RTECSNumber GL6990000
TSCA Yes
storageNote1 Air & Moisture Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only