Learn More
Copper(I) chloride, 90+%, ACS reagent
CAS: 7758-89-6 | ClCu | 99 g/mol
$419.92 - $419.92
Chemical Identifiers
| CAS | 7758-89-6 |
|---|---|
| Molecular Formula | ClCu |
| Molecular Weight (g/mol) | 99 |
| MDL Number | MFCD00010971 |
| InChI Key | OXBLHERUFWYNTN-UHFFFAOYSA-M |
| Synonym | cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech |
| PubChem CID | 62652 |
| ChEBI | CHEBI:53472 |
| IUPAC Name | chlorocopper |
| SMILES | Cl[Cu] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC405875000
|
Thermo Scientific Chemicals
405875000 |
500 g | Glass bottle |
Each for $419.92
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7758-89-6 | |
| 99 | |
| OXBLHERUFWYNTN-UHFFFAOYSA-M | |
| 62652 | |
| chlorocopper |
| ClCu | |
| MFCD00010971 | |
| cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
| CHEBI:53472 | |
| Cl[Cu] |
Specifications
| 1490.0°C | |
| 7758-89-6 | |
| Fine Crystalline Powder | |
| 90+% | |
| CuCl | |
| 01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97 | |
| cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
| OXBLHERUFWYNTN-UHFFFAOYSA-M | |
| chlorocopper | |
| 62652 | |
| 99 | |
| ACS Reagent | |
| 4.14 | |
| Copper(I) chloride, 90+% |
| 430.0°C | |
| Green | |
| 500 g | |
| ClCu | |
| MFCD00010971 | |
| 15, 2650 | |
| Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh, forming complexes, practically insoluble in alcohol and acetone | |
| Cl[Cu] | |
| 99 | |
| CHEBI:53472 | |
| ≥90% | |
| Glass bottle | |
| 4.1400g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
GHS Signal Word: Danger
EINECSNumber : 231-842-9
RTECSNumber : GL6990000
TSCA : TSCA
RUO – Research Use Only