missing translation for 'onlineSavingsMsg'
Learn More

Copper Gluconate, USP, 98-102%, Spectrum™ Chemical

Catalog No. p-7157152
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601839 500 g Poly Bottle
18601840 2.5 kg Poly Pail
18601841 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601839 Supplier Spectrum Chemical Mfg Cor Supplier No. C1334500GM
Only null left

C1334, 527-09-3, C12H22CuO14

Spectrum™ Chemical Copper Gluconate, USP is used as a nutrient and dietary supplement. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 527-09-3
Molecular Formula C12H22CuO14
Molecular Weight (g/mol) 453.84
InChI Key OCUCCJIRFHNWBP-IYEMJOQQSA-L
IUPAC Name copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Packaging Poly Bottle