missing translation for 'onlineSavingsMsg'
Learn More

Copper gauze, 50 mesh woven from 0.23mm (0.009in) dia wire

Catalog No. p-7050918
Change view
Click to view available options
Quantity:
100 x 900 mm
500 x 900 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39713WP 100 x 900 mm
AA39713WQ 500 x 900 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39713WP Supplier Thermo Scientific Chemicals Supplier No. 039713WP
Only null left

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 100 x 900 mm
Mesh Size 50 Mesh
Odor Odorless
Form Wire Cloth
Chemical Name or Material Copper gauze, Woven from 0.23mm (0.009 in.) dia. wire
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only