missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper gauze, 40 mesh woven from 0.056mm (0.0022in) dia wire
CAS: 7440-50-8 | Cu | 63.55 g/mol
Supplier: Thermo Scientific Chemicals 040907GT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Copper gauze | |
| 7440-50-8 | |
| 40 Mesh | |
| Wire Cloth | |
| MFCD00010965 | |
| RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
| copper | |
| 23978 |
| Woven from 0.056mm (0.0022 in.) dia. wire | |
| 150 x 150 mm | |
| Odorless | |
| Cu | |
| cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
| [Cu] | |
| 63.55 | |
| CHEBI:30052 |
Chemical Identifiers
| 7440-50-8 | |
| 63.55 | |
| RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
| 23978 | |
| copper |
| Cu | |
| MFCD00010965 | |
| cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
| CHEBI:30052 | |
| [Cu] |
Safety and Handling
EINECSNumber : 231-159-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only