missing translation for 'onlineSavingsMsg'
Learn More

Copper gauze, 140 mesh woven from 0.056mm (0.0022in) dia wire

Catalog No. p-7080159
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
150 x 150 mm
300 x 300 mm
75 x 75 mm
Catalog No. Quantity
AA46603FL 75 x 75 mm
AA46603GT 150 x 150 mm
AA46603HB 300 x 300 mm
3 options

Catalog No. AA46603FL

Supplier: Thermo Scientific Chemicals 046603FL

Only null left

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 75 x 75 mm
Mesh Size 140 Mesh
Odor Odorless
Form Wire Cloth
Chemical Name or Material Copper gauze, 140 mesh woven from 0.056mm (0.0022 in.) dia. wire
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only