missing translation for 'onlineSavingsMsg'
Learn More

Copper foil, 0.025mm (0.001in) thick, Puratronic™, 99.999% (metals basis)

Catalog No. AA10950HV
Change view
Click to view available options
Quantity:
25 x 200 mm
25 x 1000 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10950HV 25 x 200 mm
AA10950GZ 25 x 1000 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10950HV Supplier Thermo Scientific Chemicals Supplier No. 010950HV
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Form: approx. 1.1g/25x200mm | CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored foil
Quantity 25 x 200 mm
Odor Odorless
Vapor Pressure 0 hPa
Form Foil
Assay Percent Range 99.999% (Metals basis)
Molecular Weight (g/mol) 63.55
Chemical Name or Material Copper
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only