missing translation for 'onlineSavingsMsg'
Learn More

Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis)

Catalog No. p-7049218
Change view
Click to view available options
Quantity:
20 x 100 cm
20 x 20 cm
Catalog No. Quantity
AA13377CU 20 x 20 cm
AA13377CV 20 x 100 cm
2 options

Catalog No. AA13377CU

Supplier: Thermo Scientific Chemicals 013377CU

Only null left

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 20 x 20 cm
Odor Odorless
Weight ≈455g/20x20cm
Form Foil
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Copper foil, 1.27mm (0.05 in.) thick
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only