missing translation for 'onlineSavingsMsg'
Learn More

Copper foil, 0.675mm (0.027in) thick, annealed, 99.9% (metals basis)

Catalog No. AA40374RR
Change view
Click to view available options
Quantity:
10 x 10 cm
20 x 20 cm
20 x 100 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40374RR 10 x 10 cm
AA40374CU 20 x 20 cm
AA40374CV 20 x 100 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40374RR Supplier Thermo Scientific Chemicals Supplier No. 040374RR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 10 x 10 cm
Odor Odorless
Weight ≈241g/20x20cm
Form Foil
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Copper foil, 0.675mm (0.027 in.) thick, Annealed, Temper: soft
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only