missing translation for 'onlineSavingsMsg'
Learn More

Copper foil, 0.5mm (0.02in) thick, Puratronic™, 99.998% (metals basis), Oxygen free

Catalog No. AA36395FI
Change view
Click to view available options
Quantity:
50 x 50 mm
50 x 250 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36395FI 50 x 50 mm
AA36395GB 50 x 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36395FI Supplier Thermo Scientific Chemicals Supplier No. 036395FI

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 50 x 50 mm
Odor Odorless
Weight ≈11.2g/50x50mm
Form Foil
Percent Purity 99.998%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Copper foil, 0.5mm (0.02 in.) thick, Oxygen free
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only