missing translation for 'onlineSavingsMsg'
Learn More

Copper foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis)

Catalog No. p-7049219
Change view
Click to view available options
Quantity:
20 x 20 cm
20 x 100 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13380CU 20 x 20 cm
AA13380CV 20 x 100 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA13380CU Supplier Thermo Scientific Chemicals Supplier No. 013380CU
Only null left

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Quantity 20 x 20 cm
Odor Odorless
Weight ≈45.5g/20x20cm
Form Foil
Assay Percent Range 99.9% (metals basis)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Copper foil, 0.127mm (0.005 in.) thick, Annealed, Temper: soft
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only