missing translation for 'onlineSavingsMsg'
Learn More

Copper flake, -325 mesh, 99% (metals basis)

Catalog No. p-4408711
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4507330 250 g
AA45073A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4507330 Supplier Thermo Scientific Chemicals Supplier No. 04507330
Only null left

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Melting Point 1083°C
Boiling Point 2562°C
Appearance Copper colored flakes
Density 8.94
Quantity 250 g
Mesh Size -325 Mesh
Odor Odorless
Vapor Pressure 0 hPa
Form Flake
Assay Percent Range 99% (Metals basis)
Formula Weight 63.55
Chemical Name or Material Copper
Show More Show Less
Hazard Category H228-H319-H335-H411-H401: Flammable solid. Causes serious eye irritation. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-159-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only