missing translation for 'onlineSavingsMsg'
Learn More

Copper, EMSURE™, Fine Powder, MilliporeSigma™

Catalog No. M1027030250
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1027030250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1027030250 Supplier Technidata Supplier No. 1.02703.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recommended Storage

+5°C to +30°C.

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Melting Point 1083°C
Density 8.96 g/3 (20 °C)
Quantity 250 g
Packaging Plastic Bottle
RTECSNumber GL5325000
Recommended Storage +5°C to +30°C.