missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, 99.9%, (trace metal basis), shot 1-10 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Color | Brown-Rust |
| Melting Point | 1083.0°C |
| Boiling Point | 2595.0°C |
| Quantity | 100 g |
| Packaging | Glass bottle |
| Form | Granular |
| Assay Percent Range | 99.9% |
| Linear Formula | Cu |
| Fieser | 01,157; 02,82; 03,63; 04,102; 05,146; 07,73; 08,113; 09,122; 15,99; 16,95 |
| Solubility Information | Solubility in water: insoluble. |
| Show More |
RUO – Research Use Only