missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Copolyester, Carboy with Handle
GREENER_CHOICE

Catalog No. 19830449
Change view
Click to view available options
Capacity (English):
5.3 gal.
Capacity (Metric):
20 L
Dimensions (L x W x H):
345 x 206 x 640 mm
Closure Size:
120 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Capacity (Metric) Dimensions (L x W x H) Closure Size
19830449 5.3 gal. 20 L 345 x 206 x 640 mm 120 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19830449 Supplier Justrite™ Supplier No. 12949
Only null left

For Use With: For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.; Closure Material: Polypropylene; Color: Clear

  • Constructed from clear, BPA-free, USP Class VI, FDA grade resin
  • Rectangular ergonomic design
  • Interchangeable caps and adapters provide top connection flexibility
  • Material, volume, and metric graduation marks imprinted on ca

Specifications

Accuracy ±5%
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 120 mm
Color Clear
Dimensions (L x W x H) 345 x 206 x 640 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders. Especially when visual inspection needed.
Height (English) 25.2 in.
Length (English) 13.6 in.
Material Copolyester
Product Type Carboy with Handle
Shape Rectangular
Width (English) 8.1 in.
Show More Show Less