Learn More
Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches
Permits complete, rapid elution by keeping the entire sample close to the surface. Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches are anatomically designed swabs to optimize analyte collection efficiency with improved patient comfort.
Supplier: Copan Diagnostics CA501CS01
Description
Permits complete, rapid elution by keeping the entire sample close to the surface. Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches are anatomically designed swabs to optimize analyte collection efficiency with improved patient comfort.
Sterile, individually wrapped swabs in a plastic applicator.- Ergonomic and Anatomic Design: Improves patient comfort and specimen collection
- Rapid Automatic Elution: Instant and spontaneous release of sample into liquid media
- Measurable and Consistent Uptake and Transfer: Quantitative measurement and improved test sensitivity
- Patient Friendly: Anatomical design and a soft texture for rapidly and efficiently dislodging cells
- Compatible with Multiple Applications: Rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, bacteriology and virology culture
- Safe and Convenient: Molded breakpoint
Chemical Identifiers
| Plastic | |
| Sterile | |
| Individually-wrapped in peel pouch | |
| Specimen Collection Swab |
Specifications
| Sterile | |
| Plastic | |
| Individually-wrapped in peel pouch | |
| 3.15 in. |
| Nylon | |
| Minitip flocked | |
| 80 mm | |
| Specimen Collection Swab |