Learn More
Abcam Inc Coomassie Brilliant Blue R-250 Protein Stain, 5G
Catalog No. 502704788
Encompass
MW 825.99 g/mol. Triphenylmethane dye used in SDS-PAGE for the analysis of proteins. The dye molecule binds to proteins to form a protein-dye complex. This complex stabilizes the negatively charged anionic form of the dye producing the blue color, even under acidic conditions.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502704788 Supplier Abcam Inc Supplier No. AB1462605G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 825.99 g/mol. Triphenylmethane dye used in SDS-PAGE for the analysis of proteins. The dye molecule binds to proteins to form a protein-dye complex. This complex stabilizes the negatively charged anionic form of the dye producing the blue color, even under acidic conditions.
The product is subject to the following: Abcam Restricted Use Statement