missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue G-250, MP Biomedicals™

Catalog No. p-2366523
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19034305 5 g
ICN808274 10 g
ICN19034325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19034305 Supplier MP Biomedicals Inc Supplier No. 0219034305
Only null left

Useful dye for SDS gels as it readily stains proteins with minimal background color

  • It is a protein stain in electrophoresis.
  • It has been used in the Bradford dye-binding protein assay.
  • A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids.
  • Brilliant Blue G has been used for determining critical micelle concentration of detergents.

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]

Specifications

Synonym Acid Blue 90, Brilliant Blue G
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Molecular Weight (g/mol) 854.025
Formula Weight 854
Grade Electrophoresis
Color Blue to Purple
Quantity 5 g
Physical Form Powder
Chemical Name or Material Coomassie Brilliant Blue G-250, Protein stain in electrophoresis.