missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ V Blood Collection Tube Modules

Catalog No. 07210069
Change view
Click to view available options
Holds:
12 (13 or 16 mm Tubes)
9 (13 x 100 mm Blood Tubes or 5 mL Cryogenic Vials)
9 (13 x 75 mm Blood Tubes)
9 (16 x 100 mm Blood Tubes)
Design:
CoolRack V
CoolRack V13
CoolRack V16
CoolRack VS13
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Design
07210069 12 (13 or 16 mm Tubes) CoolRack V
07210065 9 (13 x 75 mm Blood Tubes) CoolRack VS13
07210066 9 (16 x 100 mm Blood Tubes) CoolRack V16
07210067 9 (13 x 100 mm Blood Tubes or 5 mL Cryogenic Vials) CoolRack V13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07210069 Supplier Corning™ Supplier No. 432069
Get Up to $2,500 in Products with Purchase
View all promotions

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.

  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.

Specifications

Holds 12 (13 or 16 mm Tubes)
Design CoolRack V
Type Blood Collection Tube Module