missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ Tall Modules

Catalog No. 07210068
Change view
Click to view available options
Holds:
1 250 mL Centrifuge Tube
1 250 mL Easy Grip Storage Bottle
12 x 15 mL Centrifuge Tubes
12 x 5 mL Centrifuge Tubes, SBS-compatible
4 x 50 mL Centrifuge Tubes
9 x 15 mL Centrifuge Tubes
Design:
CoolRack 15 mL
CoolRack 250 mL
CoolRack 250 mL PF
CoolRack 50 mL
CoolRack L
CoolRack XT 5 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Design
07210068 12 x 15 mL Centrifuge Tubes CoolRack L
07210060 12 x 5 mL Centrifuge Tubes, SBS-compatible CoolRack XT 5 mL
07210061 9 x 15 mL Centrifuge Tubes CoolRack 15 mL
07210062 4 x 50 mL Centrifuge Tubes CoolRack 50 mL
07210063 1 250 mL Centrifuge Tube CoolRack 250 mL PF
07210064 1 250 mL Easy Grip Storage Bottle CoolRack 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 07210068 Supplier Corning™ Supplier No. 432068
Get Up to $2,500 in Products with Purchase
View all promotions

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive centrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible

Specifications

Holds 12 x 15 mL Centrifuge Tubes
Design CoolRack L
Type Tall Module