missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ Tall Modules

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Supplier:  Corning™ 432063

 View more versions of this product

Catalog No. 07210063


Only null left

Description

Description

Corning CoolRack thermo-conductive centrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible
Specifications

Chemical Identifiers

Tall Module

Specifications

1 250 mL Centrifuge Tube
CoolRack 250 mL PF
Tall Module
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions