Learn More
Corning™ CoolRack™ SV Sample Vial Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
$1,287.78
Specifications
Type SV Sample Vial Module
View More Specs
Products 2
Catalog Number Mfr. No. Holds Design Price Quantity & Availability  
Catalog Number Mfr. No. Holds Design Price Quantity & Availability  
07210058
View Documents Promotion Details
Corning™
432058
12 x 5 mL sample vials SV2
Case for $1,287.78
Only null left
07210059
View Documents Promotion Details
Corning™
432059
12 x 10 mL sample vials SV10
Case for $1,287.78
Only null left
Description
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
  • Cell therapy injectable ampule modules
  • Eensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.
Specifications
Specifications
SV Sample Vial Module