missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ CoolRack™ Plate and Reservoir Modules
Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
$980.58 - $1,239.58
| Catalog Number | Mfr. No. | Holds | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Holds | Price | Quantity & Availability | |||||
|
07210070
|
Corning™
432070 |
1 Flat Bottom Plate, 1 SBS-compatible |
Case for $1,065.80
|
|
|||||
|
07210071
|
Corning™
432071 |
1 U-bottom Microplate, SBS compatible |
Case for $1,239.58
|
|
|||||
|
07210072
|
Corning™
432072 |
Reagent Reservoir (55 mL) |
Case for $980.58
|
|
|||||
Description
Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.
- CoolSink modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
- All CoolSink modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- All modules are compatible with all temperature sources.
- SBS-compatible
Specifications
| CoolSink Plate and Reservoir Module |