missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ 96 Storage Tube Modules

Catalog No. 07210056
Change view
Click to view available options
Holds:
96 x 0.5 mL 2D Bar Code Tubes
96 x 1.4 mL 2D Bar Code Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07210056 96 x 0.5 mL 2D Bar Code Tubes
07210057 96 x 1.4 mL 2D Bar Code Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07210056 Supplier Corning™ Supplier No. 432056

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • 2D Barcode storage tube modules
  • Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • Certain CoolRack modules are SBS-compatible.

Specifications

Holds 96 x 0.5 mL 2D Bar Code Tubes
Type Storage Tube Module