missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products Cooling Neck Shade

Catalog No. 19555021
Change view
Click to view available options
Color:
Lime
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19555021 Orange
19555020 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19555021 Supplier Pyramex Safety Products Supplier No. CNS140

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pyramex Safety™ Cooling Neck Shade keeps users heads cool and blocks sun from the neck

  • Evaporative cooling material keeps users cool for hours
  • Made from soft quilted hi-vis material for greater visibility
  • Fits on the suspension of a hard hat with hook and loop closures
  • To activate, soak in cool water for 2 to 5 minutes
  • Reusable – just reactivate

Specifications

Color Orange
For Use With (Equipment) Hard Hat
Size Universal