Learn More
Pyramex Safety Products Cooling Hard Hat Pad
Description
- Evaporative cooling material provides hours of heat stress relief
- To activate, soak in cool water for 2 to 5 minutes
- Reusable – just reactivate
Specifications
Specifications
| For Use With (Equipment) | Hard Hats |
| Size | Universal |