Learn More
Corning™ CoolCell™ LX Cell Freezing Vial Containers
Provides alcohol-free freezing at the rate of -1°C/minute when placed in a -80°C freezer.
Specifications
| Material | Polyethylene Foam |
|---|---|
| Shape | Round |
| Type | Freezer Box |
| For Use With (Application) | Cryopreservation of Most Cells and Cell Lines |
| Holds | 12 (Standard 1 to 2 mL Cryogenic Vials) |
| Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Quantity | Price | Quantity & Availability | ||||
|
07210001
|
Corning™
432001 |
Purple | 1/Pk., 1/Cs. |
N/A
|
|
||||
|
07210002
|
Corning™
432002 |
Green | 1/Pk., 1/Cs. |
N/A
|
|
||||
|
07210003
|
Corning™
432003 |
Orange | 1/Pk., 1/Cs. |
N/A
|
|
||||
|
07210004
|
Corning™
432004 |
Pink | 1/Pk., 1/Cs. |
N/A
|
|
||||
|
07202363
|
Corning™
432138 |
Assorted (Green, Orange, Pink, Purple) | 4/Pk., 1/Cs. |
N/A
|
|
||||
Description
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
- Ease of use
- Alcohol and fluid-free freezing
- Lower cost of use than alcohol-based devices
- High cell recovery and cell viability
- Reproducibility
- Simple, consistent way to standardize controlled-rate freezing
Specifications
| Polyethylene Foam | |
| Freezer Box | |
| 12 (Standard 1 to 2 mL Cryogenic Vials) | |
| 11.7 cm | |
| 12.7 mm |
| Round | |
| Cryopreservation of Most Cells and Cell Lines | |
| 11.7 x 10.11 cm | |
| 10.11 cm |