missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolCell™ 2mL Filler Vials for CoolCell™ LX and FTS30 Containers

Catalog No. 07210076
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07210076 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07210076 Supplier Corning™ Supplier No. 432076
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for repeated use. Corning™ CoolCell™ 2mL Filler Vials ensure cell freezing rate consistency and uniform results when using CoolCell containers.

Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.

Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:

  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing

Specifications

Closure Color White
Color Clear
Volume (Metric) 2 mL
Includes Glycerol
No. per Case 6
No. per Pack 6
For Use With (Equipment) CoolCell LX, CoolCell FTS30 5ml LX Containers