missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolCell™ Cell Freezing Vial Containers

Catalog No. 07210000
Change view
Click to view available options
Holds:
12 (Standard 1 to 2 mL Cryogenic Vials)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
07210000 12 (Standard 1 to 2 mL Cryogenic Vials)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07210000 Supplier Corning™ Supplier No. 432000
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Controls rate of freezing to -1°C/minute when placed in a -80°C freezer.

Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.

Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:

  • Ease of use
  • Alcohol and fluid-free freezing
  • Lower cost of use than alcohol-based devices
  • High cell recovery and cell viability
  • Reproducibility
  • Simple, consistent way to standardize controlled-rate freezing

Specifications

Description High cell recovery, cell viability
Color Purple
Material Foam
Shape Round
Type Freezer Box
Holds 12 (Standard 1 to 2 mL Cryogenic Vials)
Quantity 1/Pk., 1/Cs.
Weight (Metric) 0.19 kg