Learn More
Epredia™ Cool-Cut with universal cassette clamp
Description
- Directly fits onto the specimen cylinder of the rotary microtome line
- Ability to be equipped as a universal cassette clamp or standard specimen clamp
- Effective when extended cutting periods on the microtome are required to “step section” or “serial section” through the paraffin block
Includes:
- Peltier device with integrated specimen clamp
- Counter-balance weights
- Power supply
Specifications
Specifications
| Product Type | Standard Clamp |
| For Use With (Equipment) | Epredia Microtomes |