Learn More
ILC Dover Conversion Kit - Sentinel XL HP to XL CBRN
Description
- Comes with Butyl Hood Assembly and 3 CBRN cartridge filters
- Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
- Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
- PAPRs allow workers to move freely and unrestricted
- Sentinel XL system parts are compatible with components of the same brand and series
Specifications
Specifications
| Product Type | PAPR Conversion Kit |
| Includes | Hood Assembly and Cartridge Filters |
| Model | S-5000-05 |