missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Controlled Environment Sleeve
Description
- Microporous composite material
- Breathable and lightweight
- Low particle count
- Serged elastic at both ends
- Meets the requirements of controlled environments
Specifications
Specifications
| Product Type | Controlled Environment Sleeve |
| Size | Universal |
| Material | Microporous Fabric |
| Color | White |
| Sterility | Non-sterile |
| Disposable | Single-use |
| Closure Type | Elastic Ends with Thumb Loop |
| Seam Type | Serged |
| Latex Free | Yes |
| For Use With (Application) | Controlled and Critical Environments |
| Show More |