missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Environment Sleeve

Catalog No. 19165007
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19165007 Universal
19165007XL X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19165007 Supplier Fisherbrand™ Supplier No. MS016C158
Only null left

Microporous composite sleeves with a low particle count to meet the requirements of controlled environments and designed with serged seams for added durability and protection.

  • Microporous composite material
  • Breathable and lightweight
  • Low particle count
  • Serged elastic at both ends
  • Meets the requirements of controlled environments

Specifications

Product Type Controlled Environment Sleeve
Size Universal
Material Microporous Fabric
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends with Thumb Loop
Seam Type Serged
Latex Free Yes
For Use With (Application) Controlled and Critical Environments
Length (English) 19 in.
Length (Metric) 48.26 cm
No. per Case 150/Cs.
Cleanroom Class Class 100 (ISO 5)
Show More Show Less