missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Environment Hood Universal

Catalog No. 19165006
Catalog No. 19165006 Supplier Fisherbrand™ Supplier No. SD61A358

Fisherbrand hoods are constructed of a lightweight, high quality microporous composite material with a low particle count.

  • Microporous composite material
  • Breathable and lightweight
  • Low particle count
  • Extended fabric ties in front
  • Full range of motion and vision

Specifications

Product Type Cleanroom Hood
Size Universal
Material Microporous
Color White
Closure Type Ties
Disposable Single-use
Latex Free Yes
Seam Type Serged
Sterility Non-sterile
Packaging Type Double Bagged
Quantity 100 Pairs/Cs.
Length (English) 24 in.
Height (English) 10 in.
For Use With (Application) Controlled and Critical Environments
Resistance Particulates
Cleanroom Class Class 100 (ISO 5)
Show More Show Less