missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Environment Frock

Catalog No. p-200089306
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19165010A Small
19165010B Medium
19165010C Large
19165010D X-Large
19165010E 2X-Large
19165010F 3X-Large
19165010G 4X-Large
19165010H 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19165010A Supplier Fisherbrand™ Supplier No. FK621411
Only null left

Fisherbrand high quality microporous composite frocks are breathable, strong and lightweight.

  • Microporous composite material
  • Strong, clean and lightweight
  • Mandarin collar
  • Zipper front
  • Elastic wrist
  • Snap collar
  • Zip closure

Specifications

Product Type Controlled Environments Frock
Size Small
Material Microporous Composite Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Seam Type Serged
For Use With (Application) Controlled and Critical Environments
Collar Type Mandarin
Latex Free Yes
No. of Pockets None
Sleeve Style Raglan Sleeve
Packaging Type Double Bagged
Length (English) 40.5 in.
Cleanroom Class Class 100 (ISO 5)
Show More Show Less