missing translation for 'onlineSavingsMsg'
Learn More

CritiCore Protective Wear Controlled Environment ESD Grid Hood

Catalog No. 18111194
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18111194 3X-Large
18111192 X-Large
18111193 2X-Large
18111190 Medium
18111191 Large
18111189 Small
18111188 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18111194 Supplier CritiCore Protective Wear Supplier No. 11HD3262NANO01WHT3XL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Controlled environment ESD grid hood is reusable and made with a high-density non-linting material.

  • Tunneled elastic back plus horizontal and vertical adjustment snaps
  • Split-drape design with the front panel longer than the back
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free

Specifications

Product Type Open Face
Size 3X-Large
Material Polyester, Carbon
Color White
Closure Type Snap
Disposable Reusable
Design Type Open Face
Model 11HD3262NANO
Seam Type Pre-serged/Felled
Sterility Non-sterile
Quantity 1 Ea.
Length (English) 18.5 in.
Length (Metric) 46.99 cm
Diameter (English) 9 in.
Diameter (Metric) 22.86 cm
For Use With (Application) Micro-electronics and semi-conductor manufacturing
Show More Show Less