missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Controlled Environment Coverall
Fisherbrand microporous composite coveralls are designed for many non-hazardous applications and will keep the wearer clean and comfortable all day.
Supplier: Fisherbrand™ CV640328
Description
- Microporous composite material
- Strong and lightweight
- Tapered collar
- Zipper front
- Elastic wrist, ankle and back
- Strong stress-resistant serged seams
Specifications
| Cleanroom Coveralls | |
| Unisex | |
| White | |
| Standard | |
| Elastic | |
| Unhooded | |
| Serged | |
| Particle | |
| Elastic Back | |
| None | |
| Controlled and Critical Environments | |
| Class 100 (ISO 5) |
| 5X-Large | |
| Microporous Composite Fabric | |
| Single-use | |
| Elastic | |
| Collared | |
| Zipper | |
| Non-sterile | |
| Tapered | |
| Yes | |
| Set-in Sleeve | |
| 25/Cs. |