missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Environment Coverall with Boot Cover and Hood

Fisherbrand microporous composite coveralls with hood and boots are designed for many non-hazardous applications and will keep the wearer clean and comfortable all day. Extra coverage and protection with hood and boots.

$252.51 - $380.20

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
View More Specs

Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-165-011A
View Documents
Fisherbrand™
CV64A321
Small
Case of 25 Each for $252.51
Only null left
 
19-165-011B
View Documents
Fisherbrand™
CV64A322
Medium
Case of 25 Each for $263.90
Only null left
 
19-165-011C
View Documents
Fisherbrand™
CV64A323
Large
Case of 25 Each for $274.32
Only null left
 
19-165-011D
View Documents
Fisherbrand™
CV64A324
X-Large
Case of 25 Each for $285.07
Only null left
 
19-165-011E
View Documents
Fisherbrand™
CV64A325
2X-Large
Case of 25 Each for $296.45
Only null left
 
19-165-011F
View Documents
Fisherbrand™
CV64A326
3X-Large
Case of 25 Each for $309.40
Only null left
 
19-165-011G
View Documents
Fisherbrand™
CV64A327
4X-Large
Case of 25 Each for $322.99
Only null left
 
19-165-011H
View Documents
Fisherbrand™
CV64A328
5X-Large
Case of 25 Each for $338.16
Only null left
 
19-165-011I
View Documents
Fisherbrand™
CV64A3210
7X-Large
Case of 25 Each for $380.20
Only null left
 
Description

Description

  • Microporous composite material
  • Strong and lightweight
  • Tapered collar
  • Zipper front
  • Elastic wrist and ankle
  • Elastic hood
  • Anti-skid boots
Specifications

Specifications

Cleanroom Coveralls
Microporous Composite Fabric
Single-use
Elastic
Zipper
Non-sterile
Tapered
None
Controlled and Critical Environments
Class 100 (ISO 5)
Unisex
White
Elastic at Back
Hooded
Serged
Particle
Yes
Set-in Sleeve
25/Cs.
Videos
SDS
Documents

Documents