missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Controlled Environment Boot Cover

Catalog No. 19165013
Change view
Click to view available options
Size:
Universal
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19165013 Universal
19165013XL X-Large
191650132X 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19165013 Supplier Fisherbrand™ Supplier No. BTA1612BFS
Only null left

Fisherbrand high top anti-skid boot covers fit over the calf with an elastic top to provide a comfortable fit and to prevent the boot cover from sliding down. Sole material is clean and durable. Upper material is soft and strong for all day comfort.

  • Inherent non-skid physical properties
  • Strong enough to withstand even the most abrasive floor surfaces
  • Fabric provides barrier protection and exhibits low particle generation
  • High COF fit

Specifications

Size Universal
Material Polyethylene, Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Anti-skid
Seam Type Serged
For Use With (Application) Controlled and Critical Environments
Packaging Quantity 200/Cs.
Packaging Type Double Bagged
Length (English) 13.5 in.
Height (English) 17.5 in.
Resistance Abrasion
Product Type Controlled Environments Boot Covers
Cleanroom Class Class 100 (ISO 5)
Show More Show Less