missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Control Slides, Tissue Microarray, Breast Cancer and Normal Adjacent Tissue, 8 Matched Pairs

Catalog No. 22457506
Change view
Click to view available options
Includes:
10 Unstained, 1 Stained
24 Unstained, 1 Stained
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22457506 10 Unstained, 1 Stained
22457769 24 Unstained, 1 Stained
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22457506 Supplier Mopec™ Supplier No. SC99010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

All control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 10 contains 10 unstained slides and 1 stained slide
  • Case of 25 contains 24 unstained slides and 1 stained slide

Specifications

Coating Uncoated
Thickness (Metric) 1 mm
Includes 10 Unstained, 1 Stained
Material Glass
Species Human
Tissue Condition Cancer
Type Microarray Slide
Dimensions (LxW) 75 x 25 mm
Enhanced Surface No