missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Control Slides, Tissue Microarray, Breast Cancer and Normal Adjacent Tissue, 2 Matched Pairs

Catalog No. 22457760
Change view
Click to view available options
Control Slides, Tissue Microarray, Breast Cancer and Normal Adjacent Tissue, 2 Matched Pairs; Includes: 24 Unstained Slides, 1 Stained Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22457760 Control Slides, Tissue Microarray, Breast Cancer and Normal Adjacent Tissue, 2 Matched Pairs; Includes: 24 Unstained Slides, 1 Stained Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22457760 Supplier Mopec™ Supplier No. SC98825
Only null left

All control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.

  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 25 contains 24 unstained slides and 1 stained slide

Specifications

Coating Uncoated
Thickness (Metric) 1 mm
Includes 24 Unstained Slides, 1 Stained Slide
Material Glass
Species Human
Tissue Condition Cancer
Type Microarray Slide
Dimensions (LxW) 75 x 25 mm
Enhanced Surface No