Learn More
Mopec™ Control Slides, Special Stains, Steiner-Steiner Stain
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
$528.57 - $2806.27
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
|
22-457-458
|
Mopec™
SC82910 |
10 Unstained, 1 Stained Slide |
Case of 10 Each for $528.57
|
|
|||||
|
22-457-460
|
Mopec™
SC82920 |
20 Unstained, 2 Stained Slides |
Case of 20 Each for $913.00
|
|
|||||
|
22-457-521
|
Mopec™
SC829100 |
98 Unstained, 2 Stained Slides |
Case of 100 Each for $2,806.27
|
|
|||||
Description
- Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
- Case of 10 contains 10 unstained slides and 1 stained slide
- Case of 20 contains 20 unstained slides and 2 stained slides
- Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
| Control Slide |