Learn More
Mopec™ Control Slides, Special Stains, Periodic Acid Schiff with Digestion (PAS Diastase Stain (PAS-D))
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
$461.30 - $2383.40
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
|
22-457-790
|
Mopec™
SC82610 |
10 Unstained, 1 Stained Slide |
Case of 10 Each for $461.30
|
|
|||||
|
22-457-524
|
Mopec™
SC82620 |
20 Unstained, 2 Stained Slides |
Case of 20 Each for $759.23
|
|
|||||
|
22-457-844
|
Mopec™
SC826100 |
98 Unstained, 2 Stained Slides |
Case of 100 Each for $2,383.40
|
|
|||||
Description
- Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
- Case of 10 contains 10 unstained slides and 1 stained slide
- Case of 20 contains 20 unstained slides and 2 stained slides
- Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
| Control Slide |