Learn More
Mopec™ Control Slides, Special Stains, M. Leprae Bacteria
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
Supplier: Mopec™ SC83310
Description
- Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
- Case of 10 contains 10 unstained slides and 1 stained slide
- Case of 20 contains 20 unstained slides and 2 stained slides
- Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
| Control Slide | |
| 10 Unstained, 1 Stained Slide |